Skip to Content
Merck
All Photos(1)

Documents

05970

Sigma-Aldrich

N-Allylurea

technical, ~85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2NHCONH2
CAS Number:
Molecular Weight:
100.12
Beilstein:
1745068
EC Number:
MDL number:
UNSPSC Code:
12352103

grade

technical

Assay

~85% (HPLC)

mp

84-86 °C (lit.)

SMILES string

NC(=O)NCC=C

InChI

1S/C4H8N2O/c1-2-3-6-4(5)7/h2H,1,3H2,(H3,5,6,7)

InChI key

VPJDULFXCAQHRC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service