Skip to Content
Merck
All Photos(1)

Key Documents

00315

Sigma-Aldrich

N-(Hydroxymethyl)acetamide

≥97.0% (CHN)

Synonym(s):

Acetamidomethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHCH2OH
CAS Number:
Molecular Weight:
89.09
Beilstein:
506226
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (CHN)

mp

48-52 °C

SMILES string

CC(=O)NCO

InChI

1S/C3H7NO2/c1-3(6)4-2-5/h5H,2H2,1H3,(H,4,6)

InChI key

HWJHZLJIIWOTGZ-UHFFFAOYSA-N

Other Notes

Reagent used for chloromethylations of aromatics [avoiding the carcinogenic bis(chloromethyl) ether] and for amidomethylation reactions[1][2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.R. Maulding et al.
The Journal of Organic Chemistry, 48, 2938-2938 (1983)
F. Albericio et al.
Synthesis, 271-271 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service