Skip to Content
Merck
All Photos(1)

Key Documents

28-3809

Sigma-Aldrich

Sodium phosphate monobasic

SAJ special grade, 99.0-101.0%

Synonym(s):

Monosodium dihydrogen orthophosphate, Monosodium phosphate, Sodium dihydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaH2PO4
CAS Number:
Molecular Weight:
119.98
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

SAJ special grade

Assay

99.0-101.0%

form

powder

availability

available only in Japan

pH

4.0-4.5 (25 °C, 50 g/L)

SMILES string

[Na+].OP(O)([O-])=O

InChI

1S/Na.H3O4P/c;1-5(2,3)4/h;(H3,1,2,3,4)/q+1;/p-1

InChI key

AJPJDKMHJJGVTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Samuel Adler et al.
Gastrointestinal endoscopy, 76(6), 1170-1174 (2012-10-03)
Colon capsule endoscopy (CCE) is a noninvasive technique for the detection of colorectal lesions. However, for CCE to be offered as an out-of-clinic procedure, the system needs to automatically alert the patient when to ingest the laxative (booster). We tested
Charles W Kimbrough et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 21(20), 4576-4585 (2015-07-01)
pH-low insertion peptides (pHLIP) can serve as a targeting moiety that enables pH-sensitive probes to detect solid tumors. Using these probes in conjunction with multispectral optoacoustic tomography (MSOT) is a promising approach to improve imaging for pancreatic cancer. A pH-sensitive
Johanna Troberg et al.
Drug metabolism and disposition: the biological fate of chemicals, 43(1), 107-118 (2014-10-11)
Understanding drug glucuronidation in the dog, a preclinical animal, is important but currently poorly characterized at the level of individual enzymes. We have constructed cDNAs for the 10 dog UDP-glucuronosyltransferases of subfamily 1A (dUGT1As), expressed them in insect cells, and
Filis Kazazi-Hyseni et al.
Pharmaceutical research, 31(10), 2844-2856 (2014-05-16)
The aim of this study was the development of poly(D,L-lactide-co-glycolide) (PLGA) microspheres with controlled porosity, to obtain microspheres that afford continuous release of a macromolecular model compound (blue dextran). PLGA microspheres with a size of around 40 μm and narrow size
Samira Hosseini et al.
Biosensors & bioelectronics, 69, 257-264 (2015-03-15)
Electrospun polyhydroxybutyrate (PHB) fibers were dip-coated by polymethyl methacrylate-co-methacrylic acid, poly(MMA-co-MAA), which was synthesized in different molar ratios of the monomers via free-radical polymerization. Fabricated platfrom was employed for immobilization of the dengue antibody and subsequent detection of dengue enveloped

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service