Skip to Content
Merck
All Photos(1)

Key Documents

PRI424060129

Princeton BioMolecular Research, Inc.

2-(azepan-1-yl)-6-isopropyl-pyrimidine-4-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
263.34
MDL number:
Pricing and availability is not currently available.

Iupac Name

2-(1-azepanyl)-6-isopropyl-4-pyrimidinecarboxylic acid

Inchi Code

1S/C14H21N3O2/c1-10(2)11-9-12(13(18)19)16-14(15-11)17-7-5-3-4-6-8-17/h9-10H,3-8H2,1-2H3,(H,18,19)

InChI key

ODKKIHVSQVIULG-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

Solid

Product Link

http://store.princetonbio.com/?product_id=PBMR177967

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library