Skip to Content
Merck
All Photos(1)

Key Documents

ZMQ0ALCA1

Milli-Q

Alarm Relay Cable

For use with Milli-Q® IQ/IX systems

Synonym(s):

Alarm relay cable for Milli-Q® IQ/IX systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104214
NACRES:
JA.13

compatibility

for use with Milli-Q® IX 7003
for use with Milli-Q® IX 7005
for use with Milli-Q® IX 7010
for use with Milli-Q® IX 7015
for use with Milli-Q® IQ 7000
for use with Milli-Q® IQ 7003
for use with Milli-Q® IQ 7005
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015

Looking for similar products? Visit Product Comparison Guide

General description

Alarm relay cable allows to report alert and alarm signals from the for Milli-Q® IQ/IX systems to external control board.

Features and Benefits

This Alarm relay cable allows to keep visibility of alert and alarm signal from Milli-Q® IQ/IX systems installed in not easy accessible location.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service