Skip to Content
Merck
All Photos(1)

Key Documents

SCT086

Sigma-Aldrich

BioTracker MitoBlue Live Cell Dye

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
₩491,869

₩491,869


Estimated to ship onMay 09, 2025Details



Select a Size

Change View
1 MG
₩491,869

About This Item

UNSPSC Code:
12352207
NACRES:
NA.47

₩491,869


Estimated to ship onMay 09, 2025Details


biological source

Unknown

packaging

vial of 1 mg

manufacturer/tradename

Millipore

growth mode

N/A

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−20°C

General description

BioTracker MitoBlue Live Cell Dye is a synthetic bisbenzamidine derivative that selectively stains functional mitochondria while displaying low toxicity, bright blue emission, and high resistance to photobleaching in both live and fixed cells (1). It is a unique mitochondrial dye in that after initially staining mitochondria, the dye is transported to lysosomes in a time-dependent manner. This distinct staining pattern allows for monitoring and tracking the mitochondrial recycling mechanism in living cells (2). MitoBlue has been shown to be suitable for two-photon microscopy using a 740 nm two-photon excitation (TPE) wavelength and emission channel at 480 nm (2).

Spectral Properties</bold>
Absorbance: 329nm
Emission: 420nm

Reference:
1. Sa?nchez, Mateo I., et al. "MitoBlue: a nontoxic and photostable blue-emitting dye that selectively labels functional mitochondria." ACS chemical biology 9.12 (2014): 2742-2747.
2. Sánchez, Mateo I., et al. "MitoBlue as a tool to analyze the mitochondria-lysosome communication." Scientific reports 10.1 (2020): 1-12.

Application

Purity >97% confirmed by HPLC. Identification confirmed by HNMR, LC-MS, and elemental analysis.

Features and Benefits

Live cell imaging dye that stains functional mitochondria and tracks the mitochondrial degradation process and transport to lysosomes.

Physical form

Lyophilized. Yellow solid.

Storage and Stability

Store at -20°C, desiccate and protect from light.Note: Centrifuge vial briefly to collect contents at bottom of vial before opening

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service