Skip to Content
Merck
All Photos(2)

Key Documents

MMTUGUIDE

Millipore

MILLIFLEX OASIS® guides for electrical cable and tubing

for use with Milliflex Oasis® systems

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116129
NACRES:
NB.51

packaging

pack of 20 pieces (guides)

manufacturer/tradename

Milliflex Oasis®

application(s)

bioburden testing
food and beverages
pharmaceutical
water monitoring

compatibility

for use with Milliflex Oasis® systems

shipped in

ambient

General description

MILLIFLEX OASIS® filtration system offers a solution to simplify and secure microbial testing process with premium levels of productivity, result reliability and regulatory compliance. The MILLIFLEX OASIS® pump is a new generation vacuum pump for water and bioburden analysis in pharmaceutical environments. The guides assist with directing the pump tubing and electrical cords in the work area.

Application

MILLIFLEX OASIS® filtration pump finds its application in bioburden testing and water monitoring of beer, bottled water, cosmetics and Pharmaceutical product analysis.

Legal Information

MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service