Skip to Content
Merck
All Photos(1)

Documents

MABX2021

Sigma-Aldrich

Anti-LH (Beta-subunit) Antibody, (10mg) KC

from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

monoclonal

species reactivity

human

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

Gene Information

human ... LHB(3972)

General description

Luteinizing hormone (LH) is a two-chain glycoprotein hormone; the α-chain is identical to that of chorionic gonadotropin, TSH, and FSH, while biological specificity is attributed to the distinct β-chain. Induces ovulation, spermatogenesis, and synthesis of steroid sex hormones. LH is commonly used to monitor the timing of ovulation

Immunogen

LH Beta subunit

Application

ELISA and Lateral Flow Immunoassay.
Research Category
Signaling
Research Sub Category
Hormones & Receptors
Suggested antibody pairs MABX2021-1KC and MABX2022-1KC

Physical form

Format: Purified
Ion-exchange chromatography
Purified mouse monoclonal in buffer containing 50 mM NaCl, 10 mM PBS, pH 8.0, 0.1 % sodium azide (NaN₃).

Storage and Stability

2-8°C. For long-term storage, freeze at -20°C.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service