Skip to Content
Merck
All Photos(2)

Documents

MABS1259

Sigma-Aldrich

Anti-CYP51A1 Antibody, clone N6-P2H5*G8

clone N6-P2H5*G8, from mouse

Synonym(s):

Lanosterol 14-alpha demethylase, CYP51A1, CYPL1, CYPLI, Cytochrome P450 51A1, Cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, Cytochrome P45014DM, Cytochrome P450L1, Cytochrome P450LI, LDM, Sterol 14-alpha demethylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

N6-P2H5*G8, monoclonal

species reactivity

human

species reactivity (predicted by homology)

rat (based on 100% sequence homology), mouse (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... CYP51A1(1595)

General description

Lanosterol 14-alpha demethylase (EC 1.14.13.70; UniProt Q16850; also known as CYP51A1, CYPL1, CYPLI, Cytochrome P450 51A1, Cytochrome P450, family 51, subfamily A, polypeptide 1, Cytochrome P450-14DM, Cytochrome P45014DM, Cytochrome P450L1, Cytochrome P450LI, LDM, Sterol 14-alpha demethylase) is encoded by the CYP51A1 (also known as CYP51) gene (Gene ID 1595) in human. Cytochromes P450 (CYP) proteins are primarily membrane-associated oxidases located either in the inner membrane of mitochondria or in the endoplasmic reticulum where they function as the terminal enzymes in electron transfer chains. In addition to processing endogenous substrates, CYPs also function to metabolize exogenous drugs and potentially toxic chemicals. The human CYP superfamily consisits of 57 genes and more than 59 pseudogenes divided into 18 families and 43 subfamilies. CYP51A1 represents the only member of the CYP51 subfamily and is involved in the conversion of lanosterol to 4,4-dimethylcholesta-8(9),14,24-trien-3 -ol. Products generated by the CYP51 reaction are vital intermediates in pathways leading to the formation of cholesterol in humans, ergosterol in fungi, and other types of sterols in plants. Sterols play an important structural role in the regulation of membrane fluidity and permeability, which influence the functions of enzymes, ion channels, and other cell components embedded within.

Specificity

Clone N6-P2H5*G8 was raised against a CYP51A1 C-terminal sequence 100% conserved among human, mouse, and rat species. Target band was detected only in lysate from CYP51A1-transfected, but not from untransfected, cells (Swan, R., et al. (2016). Oncotarget. In press).

Immunogen

Ovalbumin-conjugated linear peptide corresponging to a sequence near the C-terminus of human/mouse/rat CYP51A1.

Application

Anti-CYP51A1 Antibody, clone N6-P2H5*G8, Cat. No. MABS1259, is a highly specific mouse monoclonal antibody that targets CYP51A1, validated for use in Immunohistochemistry (Paraffin) and Western Blotting.
Immunohistochemistry Analysis: Clone N6-P2H5*G8 hybridoma culture supernatant detected an upregulated CYP51A1 immunoreactivity in human colon carcinoma tissue relative to that in normal liver tissue.

Immunohistochemistry Analysis: A representative lot detected increased CYP51A1 expression in formalin-fixed, paraffin-embedded primary colorectal cancer tissue than in normal colonic mucosa. Significantly reduced CYP51A1 expression was found in lymph node metastasis compared to Dukes C (stage 3) colorectal cancer (Swan, R., et al. (2016). Oncotarget. In press).

Western Blotting Analysis: A representative lot detected a target band in lysate from CYP51A1-transfected, but not from untransfected, cells (Swan, R., et al. (2016). Oncotarget. In press).
Research Category
Signaling

Quality

Evaluated by Western Blotting in human liver microsome lysate.

Western Blotting Analysis: A 1:125 dilution of this antibody detected CYP51A1 in 50 µg of human liver microsome lysate.

Target description

~50 kDa observed. 56.81/46.31 kDa (human isoform 1/2), 56.78 kDa (mouse), and 56.71 kDa (rat) calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service