Skip to Content
Merck
All Photos(1)

Key Documents

MABN1152

Sigma-Aldrich

Anti-UBE4B, clone EPR7471, Rabbit Monoclonal Antibody

clone EPR7471, from rabbit

Synonym(s):

Ubiquitin conjugation factor E4 B, Homozygously deleted in neuroblastoma 1, Ubiquitin fusion degradation protein 2, UBE4B, HDNB1, UFD2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

antibody form

unpurified

antibody product type

primary antibodies

clone

EPR7471, monoclonal

species reactivity

mouse, human, rat

technique(s)

western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... UBE4B(10277)

General description

Ubiquitin conjugation factor E4 B, also known as Homozygously deleted in neuroblastoma 1 or Ubiquitin fusion degradation protein 2, and encoded by the gene name UBE4B or HDNB1 or UFD2, is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE4B is an additional conjugation factor, E4, which is involved in multiubiquitin chain assembly. It is also the strongest candidate in the neuroblastoma tumor suppressor.

Immunogen

Synthetic peptide corresponding to human UBE4B.

Application

Anti-UBE4B, clone EPR7471, Rabbit Monoclonal Antibody is an antibody against UBE4B for use in Western Blotting.
Produced in collaboration with Epitomics
Research Category
Neuroscience
Research Sub Category
Ubiquitin & Ubiquitin Metabolism

Quality

Evaluated by Western Blotting in HEK293, Raji, SK-OV-3, L929, L6, RAW264.7, PC12, NIH -T3, HeLa, and K562 cell lysates.

Western Blotting Analysis: A 1:1,000-10,000 dilution of this antibody detected UBE4B in HEK293, Raji, SK-OV-3, L929, L6, RAW264.7, PC12, NIH-3T3, HeLa, and K562 cell lysates.

Target description

~140 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Unpurified
Rabbit monoclonal in buffer containing PBS with up to 0.1% sodium azide, 0.05% BSA, and 50% glycerol.
Unpurified

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service