Skip to Content
Merck
All Photos(1)

Documents

MAB834-1

Sigma-Aldrich

Anti-Parainfluenza 1 Antibody, clone 251/12A

ascites fluid, clone 251/12A, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

clone

251/12A, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable

isotype

IgG2a

shipped in

wet ice

Specificity

MAB834-1 is specific for the FO/F1 fusion protein common to all parainfluenza I strains. This monoclonal exhibits no cross-reactivity to Parainfluenza II or III.

Application

Anti-Parainfluenza 1 Antibody, clone 251/12A detects level of Parainfluenza 1 & has been published & validated for use in ELISA & IF.
ELISA at1:800+

Indirect Immunofluorescence at 1:100 - 200+ (fresh frozen tissue sections)

Final working dilutions must be determined by end user.
Research Category
Infectious Diseases
Research Sub Category
Infectious Diseases - Viral

Physical form

Liquid in 0.02M PB with 0.25M NaCl, pH=7.6, 0.01% Sodium azide .

Storage and Stability

Store at +2-8 C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service