Skip to Content
Merck
All Photos(1)

Documents

MAB1334

Sigma-Aldrich

Anti-Collagen Types I, II & III Antibody, clone MMCHABC

clone MMCHABC, Chemicon®, from mouse

Synonym(s):

Interstitial Collagens

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MMCHABC, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunohistochemistry: suitable
radioimmunoassay: suitable

isotype

IgM

suitability

not suitable for Western blot

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... COL1A1(1277)

Specificity

Human collagen types I, II and III (both native and denatured). No cross reactivity with human plasma proteins, other collagen types or non-collagenous proteins in ELISA, RIA or immunoblot.

Immunogen

Human collagen type I

Application

Anti-Collagen Types I, II & III Antibody, clone MMCHABC detects level of Collagen Types I & has been published & validated for use in ELISA, RIA, IH.
ELISA: Apparent binding constant from titration curves is 0.1 nM. Can be used as capture or reporter antibody for determination of collagen I or III in sandwich ELISA, but requires combination with collagen type-specific antibody for other function.

Immunohistochemistry: cryostat sections fixed with acetone or ethanol. Staining pattern similar to polyclonal antibody to interstitial collagens. Formaldehyde fixation is not recommended.

Not suitable for use on paraffin embedded sections or immunoblots.

Optimal working dilution must be determined by the end user.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Physical form

Format: Purified
Purified from murine ascites by PEG precipitation and Sepharose CL-4B chromatography. Lyophilized Powder.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 12 months from date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Sepharose is a trademark of Cytiva

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gloria Fabris et al.
Biophysical journal, 115(9), 1770-1782 (2018-10-17)
Basement membranes (BMs) are thin layers of condensed extracellular matrix proteins serving as permeability filters, cellular anchoring sites, and barriers against cancer cell invasion. It is believed that their biomechanical properties play a crucial role in determining cellular behavior and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service