Skip to Content
Merck
All Photos(1)

Key Documents

GF424

Sigma-Aldrich

IL-23, Human

HumanKine®, recombinant, Xeno-free, >95% (SDS-PAGE), suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.75

product name

HumanKine® IL-23, Human Recombinant Xeno-Free,

Assay

>95% (SDS-PAGE)

Quality Level

feature

endotoxin free

UniProt accession no.

General description

HumanKine growth factors are expressed in a proprietary human 293 protein expression system and are manufactured without the use of any animal products and are considered Xeno-Free.
Interleukin-23 (IL-23) is a heterodimer, the combination of IL-23-specific subunit p19 and IL-12 subunit p40 comprise the biologically active protein. Like IL-12, IL-23 has been shown to activate the transcription factor STAT4 (signal transducer and activator of transcription-4). Unlike IL-12, IL-23 also seems to play a role in the signaling of memory T cells. IL-23, in conjunction with IL-17, is important in controlling the bacterial growth and inflammation response in an organism with tuberculosis. It has the ability to induce Th17 activation and stimulate inflammation in many tissue types including periodontal lesions.

Quality

Bioactivity assay: Determined by the dose-dependent secretion of h-IFN-gamma from human PBMC. ED50 is typically 2-20 ng/ml.
HumanKine IL-23 is manufactured using all non-animal reagents and is considered Xeno-Free

Physical form

Product is presented in PBS and filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt. Further dilutions should be made into appropriate buffer or medium to which protein has been added.

Legal Information

HumanKine is a registered trademark of Proteintech Group, Inc. and Humanzyme, Inc

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service