Skip to Content
Merck
All Photos(1)

Key Documents

AG349

Sigma-Aldrich

Serotonin Receptor 1A, Control Peptide for AB5406

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405

concentration

4 mg/mL

Quality Level

technique(s)

inhibition assay: suitable

UniProt accession no.

Gene Information

General description

FUNCTION: SwissProt: P08908 # This is one of the several different receptors for 5- hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.
SIZE: 422 amino acids; 46107 Da
SUBCELLULAR LOCATION: Cell membrane; Multi-pass membrane protein.
SIMILARITY: SwissProt: P08908 ## Belongs to the G-protein coupled receptor 1 family.

Application

Antibody blocking (AB5406)

Optimal working concentration must be determined by the end user.

Physical form

Liquid.

Storage and Stability

Store at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service