Skip to Content
Merck
All Photos(1)

Key Documents

8725

Millipore

OmniPur® 10X TAE Buffer, Liquid Concentrate

Synonym(s):

TRIS-Acetate-EDTA Buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
Pricing and availability is not currently available.

form

solution

color

clear colorless
haze-free

General description

Each liter of concentrate makes 10L of 1X TAE Buffer (1X = 0.04 M TRIS, 0.04 M Acetate, 0.001 M EDTA). Molecular Biology grade.

Other Notes

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service