Skip to Content
Merck
All Photos(2)

Key Documents

8.45123

Sigma-Aldrich

Diisopropyl ether (stabilised with BHT)

Msynthplus

Synonym(s):

Diisopropyl ether, 2,2-Propoxypropane, Isopropyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHOCH(CH3)2
CAS Number:
Molecular Weight:
102.17
Beilstein:
1731256
MDL number:
UNSPSC Code:
12352112
EC Index Number:
203-560-6

vapor density

3.5 (vs air)

vapor pressure

120 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

827 °F

potency

5880 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

1-21 %, 100 °F

refractive index

n20/D 1.367 (lit.)

pH

7 (20 °C)

bp

68-69 °C (lit.)

mp

−85 °C (lit.)

transition temp

flash point -28 °C

density

0.725 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC(C)OC(C)C

InChI

1S/C6H14O/c1-5(2)7-6(3)4/h5-6H,1-4H3

InChI key

ZAFNJMIOTHYJRJ-UHFFFAOYSA-N

Analysis Note

Appearance of substance (visual): clear
Colour number (Apha): ≤ 20
Assay (GC, area%): ≥ 99.0 % (a/a)
Water (K. F.): ≤ 0.10 %
Density (d 20 °C/ 4 °C): 0.722 - 0.724
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test
Ni (Nickel): lot specific result

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service