Skip to Content
Merck
All Photos(1)

Documents

8.45036

Sigma-Aldrich

Diisopropyl amine

Msynthplus

Synonym(s):

Diisopropyl amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH(CH3)2
CAS Number:
Molecular Weight:
101.19
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-558-5

vapor pressure

93.33 hPa ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

295 °C

potency

420 mg/kg LD50, oral (Rat)
2900 mg/kg LD50, skin (Rabbit)

expl. lim.

1.1-7.1 % (v/v)

pH

11.8 (20 °C, 6 g/L in H2O)

bp

83-84 °C/1013 hPa

mp

-70 °C

transition temp

flash point -13.45 °C (DIN 51755 Part 1)

density

0.72 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C6H15N/c1-5(2)7-6(3)4/h5-7H,1-4H3

InChI key

UAOMVDZJSHZZME-UHFFFAOYSA-N

Analysis Note

Appearance of substance (visual): clear
Colour number (Hazen): ≤ 20
Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.715 - 0.716
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
The analysis of residual solvents is not possible due to the physicochemical properties of the material.

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service