Skip to Content
Merck
All Photos(1)

Key Documents

8.01969

Sigma-Aldrich

1-Bromooctane

for synthesis

Synonym(s):

1-Bromooctane, Octyl bromide

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
₩127,180

₩127,180


Estimated to ship onApril 07, 2025Details



Select a Size

Change View
250 ML
₩127,180

About This Item

Linear Formula:
CH3(CH2)7Br
CAS Number:
Molecular Weight:
193.12
MDL number:
UNSPSC Code:
12352101
EC Index Number:
203-912-9

₩127,180


Estimated to ship onApril 07, 2025Details


vapor pressure

0.47 hPa ( 25 °C)

Quality Level

Assay

≥98.0% (GC)

potency

4984 mg/kg LD50, oral (Rat)
8880 mg/kg LD50, skin (Rabbit)

bp

201-202 °C/1013 mbar

mp

-55 °C

transition temp

flash point 80 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

BrCCCCCCCC

InChI

1S/C8H17Br/c1-2-3-4-5-6-7-8-9/h2-8H2,1H3

InChI key

VMKOFRJSULQZRM-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.112 - 1.114
Identity (IR): passes test

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

176.0 °F - closed cup

Flash Point(C)

80 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service