Skip to Content
Merck
All Photos(1)

Key Documents

5.33657

Sigma-Aldrich

PI4KIIIβ Inhibitor, BF738735

Synonym(s):

PI4KIIIβ Inhibitor, BF738735, 2-Fluoro-4-(2-methyl-8-(3-(methylsulfonyl)benzylamino)imidazo[1,2-a]pyrazin-3-yl)phenol, Phosphatidylinositol-phosphate-4-Kinase III β Inhibitor, BF-738735

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19FN4O3S
CAS Number:
Molecular Weight:
426.46
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

potency

5.7 nM IC50

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light gray

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

General description

A cell-permeable imidazo-pyrazine based compound that acts as a potent and reversible inhibitor of PI4KIIIβ (IC50 = 5.7 nM). Displays excellent selectivity over PI4KIIIα (IC50 = 1.7 µM) and several other lipid kinases (IC50 >10 µM). Shown to dose-dependently decrease PI4P levels, reduce oxysterol-binding protein (OSBP) localization to replication organelles (ROs) and inhibit cholesterol shuttling to ROs (at 1 µM in BGM and HeLaR19 cells transfected with CVB3) at multiplicity of infection (MOI). Exhibits uniformly potent and broad spectrum antienteroviral activity across a wide range of species (EC50 = 4-31 nM) and is inactive against RNA, DNA, and retroviruses.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PI4KIII&beta
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Strating, J.R.P.M., et al. 2015. Cell Reports.10, 600.
MacLeod, A.M., et al. 2013. ACS Med. Chem. Lett.4, 585.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service