Skip to Content
Merck
All Photos(1)

Documents

524611-M

Millipore

PIKfyve Inhibitor

The PIKfyve Inhibitor, also referenced under CAS 371942-69-7, controls the biological activity of PIKfyve. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

PIKfyve Inhibitor, 6-Amino-N-(3-(4-(4-morpholinyl)pyrido[3ʹ2ʹ:4,5]furo[3,2-d]pyrimidin-2-yl)phenyl)-3-pyridine carboxamide, PtdIns3P 5-KInase Inhibitor, YM201636

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H21N7O3
CAS Number:
Molecular Weight:
467.48
UNSPSC Code:
12352200

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 1 mg/mL

storage temp.

−20°C

InChI

1S/C25H21N7O3/c26-19-7-6-16(14-28-19)24(33)29-17-4-1-3-15(13-17)22-30-20-18-5-2-8-27-25(18)35-21(20)23(31-22)32-9-11-34-12-10-32/h1-8,13-14H,9-12H2,(H2,26,28)(H,29,33)

InChI key

YBPIBGNBHHGLEB-UHFFFAOYSA-N

General description

A cell-permeable pyridofuropyrimidine compound that selectively inhibits mammalian type III PtdInsP kinase (IC50 = 33 nM and >5 µM using murine PIKfyve and yeast Fab1, respectively), while exhibiting much reduced activity against p110α (IC50 = 3.3 µM) and little activity against type IIγ PtdInsP kinase even at concentrations as high as 10 µM. Shown to block serum-induced PtdIns(3,5)P2 production in NIH3T3 cells (80% inhibition at 800 nM) and disrupt cellular endosomal transport in a reversible manner.

Warning

Toxicity: Standard Handling (A)

Preparation Note

Enhanced solubility in DMSO can be achieved by the addition of a small drop of TFA.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Jefferies, H.B.J., et al. 2008. EMBO Reports9, 164.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service