Skip to Content
Merck
All Photos(1)

Key Documents

5.09225

Millipore

IFN-α Receptor 2 Agonist, RO8191

Synonym(s):

IFN-α Receptor 2 Agonist, RO8191, 2-(2,4- bis(Trifluoromethyl)imidazo[1,2-a][1,8]naphthyridin-8-yl)-1,3,4-oxadiazole, 8-(1,3,4-Oxadiazol-2-yl)-2,4- bis(trifluoromethyl)imidazo[1, 2-a][1, 8]naphthyridine, IFN-α R2 Agonist, Interferon-α Receptor 2 Agonist, IFN-α

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H5F6N5O
Molecular Weight:
373.21
UNSPSC Code:
12352101

Assay

≥97% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

pale yellow

solubility

DMSO: 2.5 mg/mL

storage temp.

2-8°C

General description

A non-cytotoxic (up to 10 µM and 7 d in human hepatocyte Huh-7-derived HCV replicon cultures) imidazonaphthyridine compound that directly binds to the extracelluar domain of interferon-α receptor 2 (IFNAR2) in a 1:1 stoichiometric ratio and activates cellular signaling events in a IFNAR2-dependent and IFNAR1-independent manner distinct from that of interferon-α, which stimulates cellular responses only via IFNAR1/2 heterodimerization. Cellular stimulation by RO8191 via IFNAR2 homodimerization elicits similar, but not identical, signaling events as those seen with IFN-α, IFN-β, or IFN-γ stimulation. While being a much stronger stimulator of STAT3 Y705/S727 & JAK1 Y1022/1023 phosphorylations, RO8191 is ineffective in inducing IFNAR1-dependent Tyk2 Y1054/1055 phosphorylations (1 & 10 µM; 15 min; in Huh-7 cultures). Effectively boosts host anti-HCV (EC50 = 200 nM in Huh-7-derived HCV replicon cell line) and anti-EMCV activity (90% vs. <3% viability with or without 1.23 µM RO8191 in EMCV-infected human A549 lung carcinoma cultures) by inducing ISG/IFN-stimulated genes expression both in cultures in vitro and in mice (30 mg/kg p.o.) in vivo and is efficacious in reducing serum HCV titre when administered to HCV-infected mice harboring humanized liver (30 mg/kg p.o.).
A non-cytotoxic compound that directly binds to the extracellular domain of interferon-α receptor 2 and activates signaling via receptor homodimerization.
A non-cytotoxic imidazonaphthyridine compound that directly binds interferon-α receptor 2 (IFNAR2) extracelluar domain in a 1:1 ratio and activates cellular signaling via IFNAR2 homodimerization in an IFNAR1-independent manner, eliciting similar, but not identical, signaling events as those seen with IFN--α, IFN--β, or IFN--γ stimulation. While being a much stronger stimulant of STAT3 Y705/S727 & JAK1 Y1022/1023 phosphorylations, RO8191 is ineffective in inducing IFNAR1-dependent Tyk2 Y1054/1055 phosphorylations (10 µM for 15 min; in Huh-7 cultures). Effectively boosts host anti-HCV (EC50 = 200 nM in Huh-7-derived HCV replicon cell line) and anti-EMCV (90% viability with 1.23 µM RO8191 in EMCV-infected A549 cultures) by inducing ISG/IFN-stimulated genes expression both in cultures in vitro and in mice (30 mg/kg p.o.) in vivo.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
interferon-α receptor 2 (IFNAR2)

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Konishi, H., et al. 2012. Sci. Rep.2, 259.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service