Skip to Content
Merck
All Photos(1)

Key Documents

5.04908

Sigma-Aldrich

Myeloperoxidase Inhibitor-II

Synonym(s):

Myeloperoxidase Inhibitor-II, 4-(5-Fluoro-1H-indol-3-yl)butanamide, MPO Inhibitor II

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
₩141,390

₩141,390


Estimated to ship onApril 09, 2025Details

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
10 MG
₩141,390

About This Item

Empirical Formula (Hill Notation):
C12H13FN2O
CAS Number:
Molecular Weight:
220.24
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

₩141,390


Estimated to ship onApril 09, 2025Details

New, lower price on this item!

Request a Bulk Order

Assay

≥95% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light beige

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

General description

A 5-fluorotryptamine derivative that inhibitis the MPO-catalyzed Cl-/chloride-to-OCl-/hypochlorite oxidation in the presence of H2O2 (IC50 = 18 nM by taurine/2-aminoethan-sulfonic acid chlorination assay) as well as MPO-mediated LDL ApoB-100 (ApoB100; apolipoprotein B-100) oxidation (IC50 = 18 nM by ELISA) via direct affinity binding at the myeloperoxidase active site through ϖ-ϖ interaction between its indole and the the enzyme′s heme pyrrole ring D, while exhibiting much reduced affinity toward serotonin reuptake transporter/SERT (Ki = 631 nM). Kinetic studies reveal that the inhibitor acts as a good one-electron donor for the intermediate Compound I "Fe(IV)=O...Por•+", facilitating the formation (k3 = 1,6 x 107 M-1 s-1) of Compound II (Fe(IV)-OH...Por) and thereby preventing the reduction of Compound I directly to the resting state "Fe(IV)...Por" via the "halogenation cycle", while, on the other hand, behaves as a poor electron donor for Compound II, thereby preventing the completion of the "peroxidase cycle" by stalling the further reduction of Compound II to the resting state (k4 = 92 M-1 s-1). Toxicology study using Wistar Han male rats reports no signs of adverse effects post single 10 mg/kg i.p. dosage, while signs of adverse effects are observed within the 24 h, but not after, post a higher i.p. dosage at 100 mg/kg.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
myeloperoxidase
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Soubhye, J., et al. 2013. J. Med. Chem.56, 3943.
Soubhye, J., et al. 2010. J. Med. Chem.53, 8747.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service