Skip to Content
Merck
All Photos(1)

Key Documents

480352

Sigma-Aldrich

2.5S NGF, Mouse Submaxillary Glands

Synonym(s):

2.5S-Nerve Growth Factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥95% (HPLC)

Quality Level

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

shipped in

ambient

storage temp.

2-8°C

General description

Native 2.5S nerve growth factor from mouse submaxillary glands. Enhances survival, phagocytosis, and generation of superoxide in murine neutrophils. Reported to stimulate extracellular matrix invasion by human myeloma cells.
Native 2.5S nerve growth factor from mouse submaxillary glands. Enhances the survival, phagocytosis, and generation of superoxide in murine neutrophils. Reported to stimulate extracellular matrix invasion by human myeloma cells.

Biochem/physiol Actions

ED₅₀ ≤ 15 ng/ml using a PC-12 neurite outgrowth assay

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered PBS.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 weeks at -20°C.
Reconstitute to ≥10 µg/ml in sterile H₂O or buffer containing 2% BSA.

Other Notes

Hermann, J.L., et al. 1993. Mol. Cell. Biol. 4, 1205.
Raffioni, S., and Bradshaw, R.A. 1992. Proc. Natl. Acad. Sci. USA89, 9121.
Traverse, S., et al. 1992. Biochem. J.288, 351.
Levi-Montalcini, R. 1987. In Vitro Cell. Dev. Biol.23, 227.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service