Skip to Content
Merck
All Photos(1)

Key Documents

431-50

Millipore

Centriprep Centrifugal Filter Unit

50 kDa cutoff, 15 mL volume

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
eCl@ss:
36100101

material

Nylon cap
polycarbonate collector (for filtrate)
polycarbonate reservoir (for sample)
polypropylene seal
regenerated cellulose membrane

sterility

non-sterile

feature

holdup volume 0.5 mL

manufacturer/tradename

Centriprep®

parameter

15 mL sample volume

technique(s)

protein extraction: suitable
protein purification: suitable

L

135 cm

diam.

2.81 cm

filtration area

2.84 cm2

pore size

50 kDa NMWCO

application(s)

sample preparation

shipped in

ambient

General description

Filter Code: YM-50
Filter Type: Ultrafiltration

Application

50 kDa cutoff, 15 mL volume
Swinging bucket rotor w/maximum 1,500 x g

Features and Benefits

Minimum Final Concentrate Volume: 700 µL

Physical form

Color Code: Rose

Legal Information

CENTRIPREP is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service