Skip to Content
Merck
All Photos(1)

Documents

33-04A

Sigma-Aldrich

PI 3-Kinase 3-step HTRF® Assay

Everything you need to perform the PI3 Kinase 3-step HTRF assays (1920 wells).

Synonym(s):

Kinase Activity Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

mouse, human

manufacturer/tradename

HTRF®
Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

detection method

fluorometric (HTRF)

Gene Information

human ... PIK3CA(5290)

Application

Research Category
Apoptosis & Cancer
Research Sub Category
Oncology
Used to detect/quantify: PI 3-Kinase

Biochem/physiol Actions

Target Sub-Family: PI3K/PI4K

Packaging

Kit capacity: 1920 wells

Components

4X Reaction Buffer

PIP2 (substrate, 1mM stock)

DM A Reaction Buffer

DM B Reaction Buffer

DM C Reaction Buffer

BPIP3

Stop Reagent

Storage and Stability

12 months at -70°C

Legal Information

HTRF is a registered trademark of Cis Bio International
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service