Skip to Content
Merck
All Photos(1)

Documents

219381

Sigma-Aldrich

Cathepsin K Inhibitor III

The Cathepsin K Inhibitor III controls the biological activity of Cathepsin K. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

Cathepsin K Inhibitor III, Inhibitor I, 1-(N-Benzyloxycarbonyl-leucyl)-5-(phenylalanyl-leucyl)carbohydrazide, Z-L-NHNHCONHNH-LF-NH₂

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H43N7O6
Molecular Weight:
597.71
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥97% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

DMSO: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable peptidyl bis-carbohydrazide compound that acts as a potent, selective, and reversible inhibitor of cathepsin K (Ki,app = 9.7 nM). At higher concentrations, also inhibits the activities of cathepsin L, cathepsin B, and papain (Ki,app = 120 nM, 5.1 µM, and 2.3 µM, respectively). Completely blocks cathepsin K activity in primary human synovial fibroblast cultures at 1 µM.
A cell-permeable, potent, selective, reversible inhibitor of cathepsin K (Ki,app = 9.7 nM). At higher concentrations also inhibits the activities of cathepsin L, cathepsin B, and papain (Ki,app = 120 nM, 5.1 mM, and 2.3 µM, respectively). Reported to completely block cathepsin K activity in primary human synovial fibroblast cultures at a concentration of 1 µM.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Cathepsin K
Product does not compete with ATP.
Reversible: yes
Target Ki: 9.7 nM against cathepsin K

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

H-Phe-Leu-NHNH-CO-NHNH-Leu-Z

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Wang, D., et al. 2002. Biochemistry41, 8849.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service