Skip to Content
Merck
All Photos(1)

Documents

1.14417

Supelco

Cyanide Test Kit, colorimetric

0.002-0.030 mg/L (CN⁻), MQuant®

Synonym(s):

Cyanide Visual Test Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116
NACRES:
NA.24

product name

Cyanide Test, colorimetric, 0.002-0.030 mg/L (CN-), for use with MCOLORTEST®

specific analyte(s)

cyanide

Quality Level

measuring range

0.002-0.030 mg/L (CN-)

compatibility

for use with MCOLORTEST®

detection method

colorimetric

storage temp.

15-25°C

General description

Method: colorimetric with color card and comparator block with long test tubes 0.002 - 0.004 - 0.007 - 0.010 - 0.013 - 0.016 - 0.020 - 0.025 - 0.030 mg/l CN MQuant®
The MQuant® Cyanide test (formally MColortest®) measures free cyanide ions e.g. in
  • Groundwater and surface water
  • Drinking water and mineral water
  • Waters from aquaculture
  • Industrial water
  • Wastewater and percolating water
  • Electroplating wastewater
  • Food after appropriate sample pretreatment
This test is not suited for seawater.

Method
Cyanide ions react with a chlorinating agent to form cyanogen chloride, which in turn reacts with 1,3-dimethylbarbituric acid in the presence of pyridine to form a violet dye (König reaction). The cyanide concentration is measured semiquantitatively by visual comparison of the color of the measurement solution with the color fields of a color card.
The unique brilliance of the print and their fine color graduation allow precise analyses, even at the lower end of the concentration range.
Due to the long test tubes a sensitive measurement range can be covered.
Using the refill pack 1.18457 containing all the reagents, the original test kit can be used for a very long time, making it a particularly economical and ecological system.

Legal Information

MCOLORTEST is a registered trademark of Merck KGaA, Darmstadt, Germany
MQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service