Skip to Content
Merck
All Photos(1)

Documents

05-603

Sigma-Aldrich

Anti-Luciferase Antibody, clone21

clone 21, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

21, monoclonal

species reactivity (predicted by homology)

all

manufacturer/tradename

Upstate®

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Specificity

luciferase

Immunogen

Full-length recombinant luciferase from Photinus pyralis.

Application

Detect Luciferase using this Anti-Luciferase Antibody, clone21 validated for use in WB, IH.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

routinely evaluated by immunoblot on recombinant luciferase that was added to 3T3 cell lysate

Target description

66kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Format: Purified
Protein G Chromatography

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anita S Gálvez et al.
The Journal of biological chemistry, 281(3), 1442-1448 (2005-11-18)
Up-regulation of myocardial Nix and BNip3 is associated with apoptosis in cardiac hypertrophy and ischemia, respectively. To identify mechanisms of gene regulation for these critical cardiac apoptosis effectors, the determinants of Nix and BNip3 promoter activation were elucidated by luciferase
Repression of the Arf tumor suppressor by E2F3 is required for normal cell cycle kinetics.
Aslanian, A; Iaquinta, PJ; Verona, R; Lees, JA
Genes & Development null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service