Skip to Content
Merck
All Photos(1)

Key Documents

04-1501

Sigma-Aldrich

Anti-B7-H3 (CD276) Antibody, clone MJ8

clone MJ8, from rat

Synonym(s):

B7 homolog 3, CD276 antigen, CD276 molecule, Costimulatory molecule

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
₩482,937

₩482,937


Estimated to ship onMay 12, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
₩482,937

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

₩482,937


Estimated to ship onMay 12, 2025Details


Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

MJ8, monoclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... CD276(80381)

General description

B7 homolog 3 (B7-H3 or CD276) is a costimulatory molecule belonging to the B7 family that signals through the CD28 family of molecules, including CD28, CTLA4 and ICOS. B7-H3 may be involved in the regulation of T-cell-mediated immune response and may play a protective role in tumor cells by inhibiting natural-killer mediated cell lysis, as well as a role of marker for detection of neuroblastoma cells. B7-H3 may also play a key role in providing the placenta and fetus with a suitable immunological environment throughout pregnancy. Both isoform 1 and isoform 2 appear to be redundant in their ability to modulate CD4 T-cell responses. Isoform 2 is shown to enhance the induction of cytotoxic T-cells and selectively stimulates interferon gamma production in the presence of T-cell receptor signaling. B7-H3 is ubiquitous but not detectable in peripheral blood lymphocytes or granulocytes, and it is weakly expressed in resting monocytes. B7-H3 is expressed in dendritic cells derived from monocytes, epithelial cells of sinonasal tissue, extravillous trophoblast cells and Hofbauer cells of the first trimester placenta and term placenta.

Specificity

This antibody recognizes the extracellular domain of B7-H3 (CD276).

Immunogen

Epitope: Extracellular Domain
Mouse B7-H3 Ig2a FC fusion protein, consisting of the extracellular domain linked to the Fc portion of mouse IgG2a.

Application

Detect B7-H3 (CD276) using this Anti-B7-H3 (CD276) Antibody, clone MJ8 validated for use in WB, FC, IH.
Flow Cytometry Analysis: A previous lot was used by an independent laboratory in FC (Osamu, N., et al., 2008).

Immunochistochemistry Analysis: A previous lot was used by an independent laboratory in IH (Osamu, N., et al., 2008).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

Evaluated by Western Blot in human placenta tissue lysate.

Western Blot Analysis: 0.25 µg/mL of this antibody detected BN-H3 on 10 µg of human placenta tissue lysate.

Target description

~31 kDa observed molecular weight on human lysates corresponds to isoform 2 of B7-H3.

Physical form

Format: Purified
Protein G Purified
Purified rat monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human placenta tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service