Skip to Content
Merck
All Photos(1)

Key Documents

KEY465199124

Key Organics / BIONET

1-[4-(4-tert-butylbenzoyl)-1H-pyrrol-2-yl]-2,2,2-trichloroethan-1-one

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
372.68
MDL number:

mp

228 - 230

Iupac Name

1-(4-(4-(tert-butyl)benzoyl)-1H-pyrrol-2-yl)-2,2,2-trichloroethan-1-one

Inchi Code

1S/C17H16Cl3NO2/c1-16(2,3)12-6-4-10(5-7-12)14(22)11-8-13(21-9-11)15(23)17(18,19)20/h4-9,21H,1-3H3

InChI key

NBMFYZCQYAQAJG-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/7R-0205

Melting Point

228 - 230

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/7R-0205


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library