Skip to Content
Merck
All Photos(1)

Key Documents

JWPH9999607D

J & W Pharmlab LLC

4-Hydroxy-tetrahydro-pyran-4-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
127.14

Iupac Name

4-hydroxytetrahydro-2H-pyran-4-carbonitrile

Inchi Code

1S/C6H9NO2/c7-5-6(8)1-3-9-4-2-6/h8H,1-4H2

InChI key

OXFYWRXLEMQSPC-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

off white solid

Product Link

https://www.jwpharmlab.com/home/product/03R0704

MSDS

https://www.jwpharmlab.com/home/product/03R0704


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library