Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249C0AE

J & W Pharmlab LLC

Methyl 2-bromo-3-(4-bromo-phenyl)-propionate

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
322
MDL number:

Iupac Name

methyl 2-bromo-3-(4-bromophenyl)propanoate

Inchi Code

1S/C10H10Br2O2/c1-14-10(13)9(12)6-7-2-4-8(11)5-3-7/h2-5,9H,6H2,1H3

InChI key

YRZAFKNONVRFNF-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Physical Form

Brown Liquid

Product Link

https://www.jwpharmlab.com/home/product/03R0006

MSDS

https://www.jwpharmlab.com/home/product/03R0006


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library