Skip to Content
Merck
All Photos(1)

Key Documents

COM448685059

Combi-Blocks, Inc.

3-(3-Trifluoromethoxyphenyl)phenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
254.21
MDL number:

Iupac Name

3'-(trifluoromethoxy)[1,1'-biphenyl]-3-ol

Inchi Code

1S/C13H9F3O2/c14-13(15,16)18-12-6-2-4-10(8-12)9-3-1-5-11(17)7-9/h1-8,17H

InChI key

LIMNXHCEZHSIJQ-UHFFFAOYSA-N

Purity

97%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-1835

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?YA-1835


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library