COM448658292
Combi-Blocks, Inc.
5-Iodo-2,3-dihydro-1h-inden-2-amine hydrochloride hydrate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Molecular Weight:
313.57
MDL number:
Iupac Name
5-iodo-2,3-dihydro-1H-inden-2-amine hydrochloride hydrate
Inchi Code
1S/C9H10IN.ClH.H2O/c10-8-2-1-6-4-9(11)5-7(6)3-8;;/h1-3,9H,4-5,11H2;1H;1H2
InChI key
JVOVNYUUWYMBSN-UHFFFAOYSA-N
storage temp.
Freezer
Purity
90%
Country of Origin
CN
Product Link
https://www.combi-blocks.com/cgi-bin/find.cgi?QC-4902
MSDS
https://www.combi-blocks.com/cgi-bin/find.cgi?QC-4902
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.