C12H316CF5D1
Chemcia Scientific, LLC
2-Hydroxy-4-methoxy-5-nitro-benzoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
CAS Number:
Molecular Weight:
213.15
Iupac Name
2-hydroxy-4-methoxy-5-nitrobenzoic acid
Inchi Code
1S/C8H7NO6/c1-15-7-3-6(10)4(8(11)12)2-5(7)9(13)14/h2-3,10H,1H3,(H,11,12)
InChI key
SGJMZHXWMUSUBE-UHFFFAOYSA-N
storage temp.
0-5C
Purity
95%
Country of Origin
US
Shipping Temperature
Ambient
Physical Form
Yellow Solid
Product Link
http://www.chemcia.com/product_fl02.asp?id=12187
MSDS
http://www.chemcia.com/product_fl02.asp?id=12187
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.