Skip to Content
Merck
All Photos(1)

Key Documents

B-099

Supelco

2-Butanone-2,4-DNPH

certified reference material, vial of 100 mg, Cerilliant®

Synonym(s):

2-Butanone-2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
₩489,535

₩489,535


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
₩489,535

About This Item

Empirical Formula (Hill Notation):
C10H12N4O4
CAS Number:
Molecular Weight:
252.23
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

₩489,535


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

vial of 100 mg

manufacturer/tradename

Cerilliant®

format

neat

storage temp.

−20°C

InChI

1S/C10H12N4O4/c1-3-7(2)11-12-9-5-4-8(13(15)16)6-10(9)14(17)18/h4-6,12H,3H2,1-2H3/b11-7+

InChI key

WPWSANGSIWAACK-YRNVUSSQSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service