Skip to Content
Merck
All Photos(2)

Key Documents

700074P

Avanti

T-MAS-d6

Avanti Research - A Croda Brand

Synonym(s):

110893

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
₩2,703,859

₩2,703,859


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
₩2,703,859

About This Item

Empirical Formula (Hill Notation):
C29H42D6O
CAS Number:
Molecular Weight:
418.73
UNSPSC Code:
41141804
NACRES:
NA.25

₩2,703,859


Please contact Customer Service for Availability

Request a Bulk Order

description

(4,4-dimethyl-d6)cholest-8(9),24-dien-3β-ol

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700074P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−70°C

SMILES string

C[C@]12CCC([C@](CC[C@H](O)C3(C([2H])([2H])[2H])C([2H])([2H])[2H])(C)C3CC4)=C4C1CC[C@]2([H])[C@]([H])(C)CCC=C(C)C

InChI key

CHGIKSSZNBCNDW-SWCUYMAQSA-N

Related Categories

General description

14-demethyl-lanosterol (D6) is a deuterated natural sterol.

Packaging

5 mL Amber Glass Screw Cap Vial (700074P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service