Skip to Content
Merck
All Photos(1)

Documents

ATE333216143

AstaTech, Inc.

5-PHENYLISOBENZOFURAN-1,3-DIONE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
224.22
MDL number:

Iupac Name

5-phenyl-2-benzofuran-1,3-dione

Inchi Code

1S/C14H8O3/c15-13-11-7-6-10(8-12(11)14(16)17-13)9-4-2-1-3-5-9/h1-8H

InChI key

YTRAFABYXOZRDF-UHFFFAOYSA-N

storage temp.

2-8 C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=51

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=11071


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service