Skip to Content
Merck
All Photos(1)

Key Documents

APO455837375

Apollo Scientific Ltd

2-Fluoro-5-(trifluoromethoxy)aniline

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
195.12
MDL number:

Iupac Name

2-fluoro-5-(trifluoromethoxy)phenylamine

Inchi Code

1S/C7H5F4NO/c8-5-2-1-4(3-6(5)12)13-7(9,10)11/h1-3H,12H2

InChI key

HQUWXEXDIIWBBY-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

99%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethoxyaniline

Synonym

3-Amino-alpha,alpha,alpha,4-tetrafluoroanisole

Flash Point

64-65/15mm

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/2-fluoro-5-trifluoromethoxyaniline


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library