Z548839
Aldrich® solvent-pouring funnel
stem L × O.D. 100 mm × 28 mm
All Photos(1)
About This Item
Recommended Products
stem L × O.D.
100 mm × 28 mm
Related Categories
General description
Unique, closed-end glass funnel has 2 mm diameter holes in the stem permitting the gentle addition of solvents to chromatography columns without disturbing the silica gel/sand layer. The top of the funnel is 75 mm in diameter.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC