Skip to Content
Merck
All Photos(1)

Documents

Z222992

Sigma-Aldrich

Stainless steel desiccator cabinet

size large

Synonym(s):

Desiccator Cabinet, Stainless Steel Cabinet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

size

large

W × L × H

41.9 cm × 58.4 cm × 57.2 cm

chamber W × L × H

40 cm × 50.2 cm × 55.2 cm

Looking for similar products? Visit Product Comparison Guide

General description

Satin finish with triple-action bar lock; door and back wall of 1/4 in. glass.
Shelf size 48.3 x 34.3 cm
Type 302 SS with glass door and walls and two expanded SS shelves with 1/2 in. adjustment. One-piece neoprene gasket seal and a full-length door hinge for an air-tight, moisture-proof closure. Shelves are adjustable and come in 2 different sizes. Manual pressure/vacuum relief valve and SS desiccant tray. No vacuum or pressure should be applied.


Applications:
  • Storing reagents, IR cells or soil samples
  • Drying biological specimens
  • Cooling crucibles
  • Storage sludge samples

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service