Key Documents
WHA77700062
Whatman® UNIFILTER® 96 well filter plates
0.45 μm, clear polystyrene, non-sterile, lysate clarification, 25 ea
Synonym(s):
Z747041, micro plate, multiwell plate, well plates
About This Item
UNSPSC Code:
41104900
NACRES:
NB.24
Pricing and availability is not currently available. Contact Technical Service
Product Name
Whatman® UNIFILTER® plates, 96 well, Lysate Clarification, well maximum volume 800 μL, lysate clarification media and 0.45?m polypropylene membrane
material
polypropylene
polystyrene membrane (rigid high grade)
sterility
non-sterile
feature
lysate clarification media and 0.45?m polypropylene membrane
packaging
pkg of 25 ea
manufacturer/tradename
Whatman 7770-0062
Whatman Article No. 28420978 (US reference)
well maximum volume
800 μL
pore size
0.45 μm
binding type
non-treated surface
Features and Benefits
  • No cross talk. Patented integral filter design prevents well-to-well cross contamination.
  • Economical to use. Wide range of well volume options ensures efficient use of materials.
  • Better control. Choice of filter media allows control of the flow rates and retention characteristics.
  • Versatile. A broad range of filtration media is available, including glass fiber, polypropylene, cellulose nitrate, cellulose acetate, nylon, and ion exchange cellulose.
Other Notes
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Legal Information
UNIFILTER is a registered trademark of Cytiva
Whatman is a registered trademark of Cytiva
Choose from one of the most recent versions:
Certificates of Analysis (COA)