Skip to Content
Merck
All Photos(1)

Documents

SLW1526/22D

Pyrex® Reagent bottles, unground narrow neck

capacity 20000 mL

Synonym(s):

Pyrex, SciLabware 1526/22D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

Pyrex bottle
colorless bottle
narrow-mouth bottle

Joint

Joints ST/NS neck (59)

feature

without cap

packaging

pack of 1 ea

manufacturer/tradename

SciLabware 1526/22D

bottle diam. × H

288 mm × 492 mm

capacity

20000 mL

neck I.D.

59 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Complies with ISO 4796
  • Borosilicate glass with unground neck
  • For use with rubber stoppers (not supplied)

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service