Skip to Content
Merck
All Photos(1)

Key Documents

GF83317349

Stainless Steel - AISI 302 alloy, FeCr18Ni8

foil, 25 x 25mm, 0.2mm thickness, hard

Synonym(s):

FE200297

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
₩195,489
5 EA
₩791,599
10 EA
₩1,446,375

₩195,489


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

Select a Size

Change View
1 EA
₩195,489
5 EA
₩791,599
10 EA
₩1,446,375

About This Item

UNSPSC Code:
11171600
NACRES:
NA.23

₩195,489


Estimated to ship onApril 11, 2025Details


Request a Bulk Order

form

foil (temper hard)

manufacturer/tradename

Goodfellow 833-173-49

size × thickness

25 mm × 25 mm × 0.2 mm

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service