GF59147523
Monel® alloy 400, Ni65Cu33Fe2
foil, 15mm disks, 0.12mm thickness, annealed
Synonym(s):
NI110350
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
form
annealed
foil (disks)
manufacturer/tradename
Goodfellow 591-475-23
size × thickness
15 mm × 0.12 mm
Looking for similar products? Visit Product Comparison Guide
General description
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Legal Information
Monel is a registered trademark of Special Metals Corporation
Monel is a registered trademark of Huntington Alloys Corp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service