Skip to Content
Merck
All Photos(1)

Documents

DWK213546107

Duran® Powder Funnel, Short Wide Stem

short, wide mouth stem(s)

Synonym(s):

213546107

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121809
NACRES:
NB.43

material

short, wide mouth stem(s)

packaging

pkg of 1 ea

manufacturer/tradename

DURAN® 213546107

funnel diam.

200 mm

Looking for similar products? Visit Product Comparison Guide

Application

Transfering of powdered substances and granulated material,The spillage of substances is avoided, Transfering, Boro 3.3

Features and Benefits

With short and wide stem; Large stem diameter makes it suitable for transferring solids; Fired-on, highly durable white print

Legal Information

DURAN is a registered trademark of DWK Life Sciences

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service