Skip to Content
Merck
All Photos(1)

Key Documents

CLS7666100

Pyrex® snap cap

snap cap size 100

Synonym(s):

polyethylene cap, polyethylene caps, stopper, Corning® 7666

Sign Into View Organizational & Contract Pricing

Select a Size

6 EA
₩22,127

₩22,127


Please contact Customer Service for Availability


Select a Size

Change View
6 EA
₩22,127

About This Item

UNSPSC Code:
41122500
NACRES:
NB.43

₩22,127


Please contact Customer Service for Availability

material

low-density polyethylene

snap cap size

100

manufacturer/tradename

Corning 7666-100

color

blue

Looking for similar products? Visit Product Comparison Guide

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service