Skip to Content
Merck
All Photos(1)

Documents

CLS7095B9

Corning® Pasteur pipettes, non-sterile

L 9 in. (228 mm), standard tip, soda lime

Synonym(s):

pipettes, serological pipets, serological pipettes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500
eCl@ss:
32079210
NACRES:
NB.04

material

soda-lime glass

sterility

non-sterile

feature

graduations: no
subdivision: no

packaging

case of 1000 ea
pack of 200 ea

manufacturer/tradename

Corning 7095B-9

L

9 in. (228 mm)

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® disposable Pasteur pipettes are made of sturdy soda-lime glass.
  • Single-use pipettes prevent the risk of cross-contamination of valuable specimens or laboratory reagents.
  • Feature a constriction one inch below the top to facilitate plugging.
  • Ideal for use in non-volumetric transfer work in bacteriology, immunology, hematology, and serology studies, as well as blood bank and clinical chemistry procedures.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service