Skip to Content
Merck
All Photos(1)

Key Documents

CLS431510

Pyrex® pear-shaped flask, with ST 14/20 outer joint

capacity 10 mL

Synonym(s):

Corning® 4315

Sign Into View Organizational & Contract Pricing

Select a Size

12 EA
₩373,013
4 EA
₩1,287,325

₩373,013


Please contact Customer Service for Availability


Select a Size

Change View
12 EA
₩373,013
4 EA
₩1,287,325

About This Item

UNSPSC Code:
41121804
NACRES:
NB.43

₩373,013


Please contact Customer Service for Availability

material

borosilicate glass
pear-shaped flask
short

sterility

non-sterile

feature

closure type Standard Taper Joint

packaging

case of 12
pack of 2

manufacturer/tradename

Corning 4315-10

capacity

10 mL

neck size

(1)

tube size

10 mL

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Pear-shaped boiling flasks are made up of borosilicate glass
  • For greater shock resistance, carefully controlled wall thicknesses are used to provide the ideal balance between mechanical strength and thermal expansion
  • Under the toughest lab conditions, their exceptional chemical durability ensures contamination-free results and long life
  • Compatible with a wide range of combinations for various assemblies
  • Contains no animal-derived materials
  • Product is not intended for use in vacuum or pressure applications

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service