C48805
2-Chloro-4-methylquinoline
99%
Synonym(s):
2-Chlorolepidine, 2-Chloro-4-methylquinoline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C10H8ClN
CAS Number:
Molecular Weight:
177.63
Beilstein:
118333
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
99%
form
powder
bp
296 °C (lit.)
mp
55-58 °C (lit.)
SMILES string
Cc1cc(Cl)nc2ccccc12
InChI
1S/C10H8ClN/c1-7-6-10(11)12-9-5-3-2-4-8(7)9/h2-6H,1H3
InChI key
PFEIMKNQOIFKSW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service